Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-1710

Sigma-Aldrich

2-Methoxyethanol

JIS special grade, ≥99.0%

Synonym(s):

Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

JIS special grade

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

548 °F

expl. lim.

24.5 %

availability

available only in Japan

refractive index

n20/D 1.402 (lit.)

pH

5.0-7.0 (25 °C)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makoto Takei et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(2), 643-652 (2010-07-10)
Ethylene glycol monomethyl ether (EGME) is a widely used industrial solvent known to cause adverse effects to human and other mammals. Organs with high metabolism and rapid cell division, such as testes, are especially sensitive to its actions. In order
Anna Maria Piras et al.
International journal of pharmaceutics, 357(1-2), 260-271 (2008-03-04)
The preparation of novel biocompatible polymeric nanoconstructs suitable to load sensitive bioactive protein agents is reported. Nanoparticles were prepared as based on hybrid polymeric matrices consisting of synthetic bioerodible alternating copolymers of maleic anhydride and n-butylvinylether hemiesterified with 2-methoxyethanol and
Frank Welsch
Toxicology letters, 156(1), 13-28 (2005-02-12)
Numerous experimental studies have established that only a few among the large family of ethylene glycol ethers (EGEs) elicit toxicity on reproduction in either gender. Notable are the monomethyl (EGME) and monoethyl (EGEE) ethers and their respective acetate esters whose
Alberto Dessy et al.
Colloids and surfaces. B, Biointerfaces, 87(2), 236-242 (2011-06-17)
Therapeutic properties of Dead Sea Water (DSW) in the treatment of skin diseases such as atopic dermatitis, psoriasis and photo aging UV damaged skin have been well established. DSW is in fact rich in minerals such as calcium, magnesium, sodium
H-Y Chang et al.
Occupational and environmental medicine, 61(8), 697-702 (2004-07-20)
To evaluate the protective effectiveness of gloves from occupational exposure to 2-methoxyethanol (2-ME); and to examine the association of 2-methoxyacetic acid (MAA) in urine and plasma collected simultaneously from low 2-ME exposure and high 2-ME exposure workers in a semiconductor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service