Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-1625

Sigma-Aldrich

1,2-Dimethoxyethane

SAJ first grade, ≥99.0%

Synonym(s):

mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1209237
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.1 (20 °C, vs air)

vapor pressure

48 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

396 °F

expl. lim.

10.4 %

availability

available only in Japan

refractive index

n20/D 1.379 (lit.)

pH

~7

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Martins et al.
Organic letters, 12(22), 5186-5188 (2010-10-19)
A palladium-catalyzed crossed biaryl coupling/reduction sequence enables the formation of meta-substituted biaryls via solvent-mediated arylpalladium(II) reduction. Isotope labeling studies determined that the decomposition of 1,2-dimethoxyethane (DME) is indeed involved in the reductive process.
Peter A Campochiaro et al.
Ophthalmology, 122(3), 545-554 (2014-12-03)
AKB-9778 is a small-molecule competitive inhibitor of vascular endothelial-protein tyrosine phosphatase (VE-PTP) that promotes Tie2 activation and reduces vascular leakage and neovascularization in mouse models. The purpose of this study was to test the safety, tolerability, pharmacokinetics, and biological activity
Guangmin Zhou et al.
Nature communications, 6, 7760-7760 (2015-07-18)
Lithium-sulphur batteries with a high theoretical energy density are regarded as promising energy storage devices for electric vehicles and large-scale electricity storage. However, the low active material utilization, low sulphur loading and poor cycling stability restrict their practical applications. Herein
Luuk M van Langen et al.
Organic letters, 7(2), 327-329 (2005-01-14)
[Reaction: see text] The (R)-oxynitrilase from almonds was immobilized as a cross-linked enzyme aggregate (CLEA) via precipitation with 1,2-dimethoxyethane and subsequent cross-linking using glutaraldehyde. The resulting preparation was a highly effective hydrocyanation catalyst under microaqueous conditions, which suppress the nonenzymatic
Jan M Englert et al.
Nature chemistry, 3(4), 279-286 (2011-03-25)
Graphene, a truly two-dimensional and fully π-conjugated honeycomb carbon network, is currently evolving into the most promising successor to silicon in micro- and nanoelectronic applications. However, its wider application is impeded by the difficulties in opening a bandgap in its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service