Skip to Content
MilliporeSigma

09-0832

Ethyl alcohol, pure

≥99.5%, for residue analysis, JIS 1000

Synonym(s):

Ethyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
PubChem Substance ID:
UNSPSC Code:
15101802
EC Number:
200-578-6
MDL number:
Beilstein/REAXYS Number:
1718733
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis

agency

JIS 1000

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

assay

≥99.5%

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

availability

available only in Japan

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

suitability

suitable for residue analysis

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Other Notes

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration


Still not finding the right product?

Explore all of our products under Ethanol


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library