Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-0590

Sigma-Aldrich

Ethanolamine

JIS special grade, ≥99.0%

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

JIS special grade

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

1436 °F

expl. lim.

17 %

availability

available only in Japan

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Vance
Biochimica et biophysica acta, 1045(2), 128-134 (1990-07-16)
Monolayer cultures of rat hepatocytes have been examined for their ability to secrete ethanolamine plasmalogen as a component of nascent lipoproteins. In culture medium from these cells, ethanolamine plasmalogen comprises approx. 20-30% of total ethanolamine glycerophospholipids when measured either as
H Perschak et al.
Human neurobiology, 6(3), 191-194 (1987-01-01)
Concentrations of putative neuroactive substances glutamate, aspartate, gamma-aminobutyric acid, glycine, proline and ethanolamine were determined in ventricular cerebrospinal fluid collected in patients suffering from Parkinson's disease, pain syndromes or cerebellar tremor. Values are similar to those given in the literature
Ye-Jin Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(31), 4578-4584 (2015-07-03)
By controlling the polymer/polymer blend self-organization rate, all-polymer solar cells composed of a high-mobility, crystalline, naphthalene diimide-selenophene copolymer acceptor and a benzodithiophene-thieno[3,4-b]thiophene copolymer donor are achieved with a record 7.7% power conversion efficiency and a record short-circuit current density (18.8
Danielle A Garsin
Nature reviews. Microbiology, 8(4), 290-295 (2010-03-18)
Ethanolamine is a compound that can be readily derived from cell membranes and that some bacteria can use as a source of carbon and/or nitrogen. The complex biology and chemistry of this process has been under investigation since the 1970s
Gabriel da Silva
The journal of physical chemistry. A, 116(45), 10980-10986 (2012-09-22)
The alkanolamine 2-aminoethanol (NH(2)CH(2)CH(2)OH), otherwise known as monoethanolamine (MEA), is a widely used solvent for carbon capture, yet relatively little is known about its atmospheric chemistry. The hydroxyl radical initiated oxidation of MEA is thought to predominantly form the α-aminoalkyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service