Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-0090

Sigma-Aldrich

Decane

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ special grade

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

410 °F

expl. lim.

2.6 %

availability

available only in Japan

refractive index

n20/D 1.411 (lit.)

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Sotthewes et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3662-3667 (2013-02-21)
The energetics and dynamics of the various phases of decanethiolate self-assembled monolayers on Au(111) surfaces were studied with scanning tunneling microscopy. We have observed five different phases of the decanethiolate monolayer on Au(111): four ordered phases (β, δ, χ*, and
Ilya V Pobelov et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(16), 164210-164210 (2012-04-03)
We describe a new setup for simultaneous measurements of force and current in conductive nanocontacts in a liquid environment with a high sampling rate and resolution. A lab-built current-to-voltage converter allows measurements of the current over seven orders of magnitude.
Ching-Wei Njauw et al.
Langmuir : the ACS journal of surfaces and colloids, 29(12), 3879-3888 (2013-02-28)
It has been known that the addition of bile salts to lecithin organosols induces the formation of reverse wormlike micelles and that the worms are similar to long polymer chains that entangle each other to form viscoelastic solutions. In this
Elena Stanoeva et al.
Bioorganic & medicinal chemistry, 13(1), 17-28 (2004-12-08)
The 2,9,10-trioxatricyclo[4.3.1.0(3,8)]decane moiety is a tetracyclic cage-like orthoester incorporated in the structure of a series of daphnane derivatives such as resiniferatoxin (RTX), kirkinine, synaptolepis factors, huratoxin etc., exhibiting various biological activities. The approaches to the preparation of cage-like orthoesters starting
Alexandra Nemkevich et al.
Physical chemistry chemical physics : PCCP, 12(45), 14916-14929 (2010-10-15)
A set of model compounds covering a range of polarity and flexibility have been simulated using GAFF, CHARMM22, OPLS and MM3 force fields to examine how well classical molecular dynamics simulations can reproduce structural and dynamic aspects of organic molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service