Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-5060

Sigma-Aldrich

Ammonium sulfamate

JIS special grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SO3NH2
CAS Number:
Molecular Weight:
114.12
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.5%

form

crystalline

availability

available only in Japan

pH

4.5-6.0 (25 °C, 114.1 g/L)

mp

131-135 °C (lit.)

SMILES string

N.NS(O)(=O)=O

InChI

1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3

InChI key

GEHMBYLTCISYNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dengfeng Dou et al.
Bioorganic & medicinal chemistry, 19(20), 5975-5983 (2011-09-20)
A new class of compounds that exhibit anti-norovirus activity in a cell-based system and embody in their structure a cyclosulfamide scaffold has been identified. The structure of the initial hit (compound 2a, ED(50) 4 μM, TD(50) 50 μM) has been
Cobalt-catalyzed direct arylation and benzylation by C-H/C-O cleavage with sulfamates, carbamates, and phosphates.
Weifeng Song et al.
Angewandte Chemie (International ed. in English), 51(33), 8251-8254 (2012-07-14)
Christopher S Adams et al.
Journal of the American Chemical Society, 134(26), 10807-10810 (2012-06-20)
Nitrogen-containing stereotriads, compounds with three adjacent stereodefined carbons, are commonly found in biologically important molecules. However, the preparation of molecules bearing these motifs can be challenging. Herein, we describe a modular oxidation protocol which converts a substituted allene to a
Laszlo Tarko et al.
Bioorganic & medicinal chemistry, 21(6), 1404-1409 (2012-12-05)
The last version of the PRECLAV algorithm was used to investigate a series of sulfamate/sulfamide carbonic anhydrase (CA, EC 4.2.1.1) inhibitors. PRECLAV allows identification of outliers for lead hopping, significant molecular fragments and similarity computation of a calibration set vs.
Jean-Yves Winum et al.
Bioorganic & medicinal chemistry, 21(6), 1419-1426 (2012-12-04)
Targeting tumour associated carbonic anhydrase (CA, EC 4.2.1.1) isoforms IX and XII is now considered as a pertinent approach for the development of new cancer therapeutics against hypoxic tumours. In the last period, with the help of X-ray crystallography, much

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service