Skip to Content
MilliporeSigma

01-4730

Ammonium formate

SAJ first grade, ≥95.0%

Synonym(s):

Formic acid ammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
EC Number:
208-753-9
UNSPSC Code:
12352107
PubChem Substance ID:
Beilstein/REAXYS Number:
3625095
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

SAJ first grade

assay

≥95.0%

form

crystalline

availability

available only in Japan

pH

5.5-7.5 (25 °C, 63.1 g/L)

mp

119-121 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Ammonium formate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library