Skip to Content

GPT-RO

Roche

Glutamate-Pyruvate Transaminase (GPT)

from pig heart

Synonym(s):

alanine aminotransferase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
Specific activity:
≥80 units/mg protein (At 25 °C with L-alanine and 2-oxoglutarate as the substrates.)
Biological source:
pig heart
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

pig heart

form

suspension

specific activity

≥80 units/mg protein (At 25 °C with L-alanine and 2-oxoglutarate as the substrates.)

packaging

pkg of 1 mL (10105589001 [10 mg]), pkg of 2.5 mL (10737127001 [25 mg])

manufacturer/tradename

Roche

optimum pH

~8.0

shipped in

wet ice

storage temp.

2-8°C

General description

Glutamate-pyruvate transaminase is a blood enzyme. It metabolizes glutamate in blood resulting in low extracellular levels of brain glutamate.

Application

Synthesis of unnatural L-amino acids from α-keto acids.

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Preparation Note

Activator: Pyridoxalphosphate
Storage conditions (working solution): The enzyme is stable when dissolved in human serum and frozen in aliquots. For best results, centrifuge the enzyme and decant the supernatant before dissolving the enzyme in human serum to avoid high ammonium sulfate concentrations in the solution.

Analysis Note

Contaminants: < 0.01% GIDH, LDH, and MDH each, < 0.03% GOT

Other Notes

For life science research only. Not for use in diagnostic procedures.
Unit Conversion: 1 U [+25 °C, pH 7.5] ≈ 1.75 U [+37 °C, pH 7.5]


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library