Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CHLUM-RO

Roche

BM Chemiluminescence Western Blotting Substrate (POD)

pkg of 1 set (11500708001 [1,000 cm2 membrane]), pkg of 1 set (11500694001 [4,000 cm2 membrane])

Synonym(s):

POD, Western blot

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332

packaging

pkg of 1 set (11500694001 [4,000 cm2 membrane])
pkg of 1 set (11500708001 [1,000 cm2 membrane])

Quality Level

manufacturer/tradename

Roche

storage temp.

2-8°C

Application

BM chemiluminescence western blotting substrate (POD) has been used along with horseradish peroxidase-coupled secondary antibodies, to visualize cells on the membranes.

Features and Benefits

Sensitivity: <10pg antigen in western blots. Shows at least 10-fold higher sensitivity than other nonradioactive or radioactive detection systems on PVDF membranes (using β-galactosidase, anti-β-galactosidase, or anti-mouse IgG-POD, Fab fragments).

Packaging

1 kit containing 3 components

Quality

Each lot is function tested in a dot blot.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Luminol Substrate Solution with enhancer

  • NaBO3 Starting Solution

  • Blocking Reagent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kinome-wide functional analysis highlights the role of cytoskeletal remodeling in somatic cell reprogramming.
Sakurai K, et al.
Cell Stem Cell, 523-34 null
Characterization of the platelet granule proteome: Evidence of the presence of MHC1 in alpha-granules
Zufferey A, et al.
Journal of Proteomics, 130-40 null
Syncytin proteins incorporated in placenta exosomes are important for cell uptake and show variation in abundance in serum exosomes from patients with preeclampsia.
Vargas A, et al.
Faseb Journal, 3703-19 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service