Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

11427857910

Roche

Fluorescein-12-UTP

≥85% (HPLC), solution

Synonym(s):

Fluorescein-12-UTP tetralithium salt, Fluorescein-5(6)-carboxamidocaproyl-(5-[3-aminoallyl]uridine 5′-triphosphate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H41N4O22P3
CAS Number:
Molecular Weight:
1010.68
MDL number:
UNSPSC Code:
41116100

Quality Level

Assay

≥85% (HPLC)

form

solution

mol wt

1034.4

packaging

pkg of 25 μL (250 nmol; 10 mM)

manufacturer/tradename

Roche

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Quality
Typical analysis: At least 85% Fluorescein-12-UTP (HPLC, area%).
Fluorescein is bound to uridine triphosphate via an amide linkage. Molecular formula is C39H37N4O22P3Li4.

Application

Fluorescein-12-UTP is used as a substrate for SP6, T3, and T7 RNA polymerases. It can replace UTP in the in vitro transcription reaction for RNA labeling.
Labeled RNA can be subsequently detected by in situ hybridization and direct fluorescence detection or detection by ELISA using Anti-Fluorescein-AP, Fab fragments or Anti-Fluorescein-POD, Fab fragments.

Other Notes

For life science research only. Not for use in diagnostic procedures.
For your convenience, fluorescein-UTP is also available in a convenient labeling mix (single solution with conventional NTPs and Fluorescein-12-UTP).

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Yu Zhang et al.
Scientific reports, 10(1), 7429-7429 (2020-05-06)
Bioinformatic analysis reveals an enrichment of putative DNA:RNA hybrid G-quadruplex-forming sequences (PHQS) on both sides of the transcription start sites (TSSs) in the genome of warm-blooded animals, suggesting a positive selection of PHQSs in evolution and functional role of DNA:RNA
Gilbert Lauter et al.
Neural development, 6, 10-10 (2011-04-07)
In recent years, mapping of overlapping and abutting regulatory gene expression domains by chromogenic two-color in situ hybridization has helped define molecular subdivisions of the developing vertebrate brain and shed light on its basic organization. Despite the benefits of this
Ryudo Ohbayashi et al.
Bio-protocol, 8(15), e2958-e2958 (2018-08-05)
Cyanobacteria are prokaryotic organisms that carry out oxygenic photosynthesis. The fresh water cyanobacterium Synechococcus elongatus PCC 7942 is a model organism for the study of photosynthesis and gene regulation, and for biotechnological applications. Besides several freshwater cyanobacteria, S. elongatus 7942

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service