Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10602361001

Roche

Plasmin, human

from human plasma

Synonym(s):

Plasmin

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

human plasma

Quality Level

Assay

95% (electrophoresis)

form

suspension

specific activity

~8 units/mg protein (At 25 °C with Chromozym PL as the substrate.)

mol wt

Mr ~85 kDa

packaging

pkg of 0.5 mL (5 U)

manufacturer/tradename

Roche

optimum pH

8.9

shipped in

wet ice

General description

Inhibitors: DFP, PMSF, TLCK, leupeptin, aprotinin, soybean trypsin inhibitor, α1-antitrypsin, α2-macroglobulin, anti-thrombin III heparin, APMSF, and α2-antiplasmin
Fibrinolysin

Specificity

Serine endopeptidase that hydrolyzes peptide and ester bonds at the carboxylic side of Lys or Arg; more selective than trypsin.

Application

Human plasmin has been used as an oligopeptide substrate in peptidase inhibition assays.

Biochem/physiol Actions

Plasmin is a direct acting fibrinolytic enzyme present in blood. Its activity is independent of plasminogen presence during fibrinolysis. Plasmin is rapidly and irreversibly inhibited by α-2 antiplasmin.

Physical form

Suspension in 3.2 M ammonium sulfate solution

Preparation Note

Stabilizers: Human plasmin does not contain stabilizers.
Storage conditions (working solution): 2 to 8 °C
Diluted solution (pH 8.6) is stable at 2 to 8 °C for at least 1–2 weeks. For storage of dialyzed solution freezing in aliquots is recommended.

Storage and Stability

Store at 2 to 8 °C. (Recommended form for storage is ammonium sulfate suspension 3.2 M, pH 8.6.)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and partial characterization of a novel serpin from Eudiplozoon nipponicum (Monogenea, Polyopisthocotylea
Roudnicky P, et al.
Parasite (Paris, France) (2018)
A novel Kunitz protein with proposed dual function from Eudiplozoon nipponicum (Monogenea) impairs haemostasis and action of complement in vitro
Jedlikova L, et al.
International Journal For Parasitology (2019)
R D Shlansky-Goldberg et al.
Journal of thrombosis and haemostasis : JTH, 6(6), 944-950 (2008-04-04)
Hemodialysis (HD) grafts often fail because of stenosis at the venous anastomosis and thrombotic occlusion. Percutaneous management relies on thrombolysis with plasminogen activators, mechanical removal of thrombus, and angioplasty of the stenotic lesion. This report describes a phase I trial
Jialu Liu et al.
NPJ Regenerative medicine, 7(1), 34-34 (2022-07-01)
The biomaterial fibrin is widely used as a clinical tissue sealant in surgery. In preclinical research, fibrin is also extensively studied as a carrier material for growth factor delivery. In these applications, premature fibrin degradation leads to recurrent bleeding, tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service