Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10378445001

Roche

Chromozym PK

90% (enzymatic), powder, pkg of 20 mg

Synonym(s):

C33H39N8O6COOCH3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

description

Molecular Formula: C33H39N8O6COOCH3

Quality Level

Assay

90% (enzymatic)

form

powder

mol wt

Mr 702.9

packaging

pkg of 20 mg

manufacturer/tradename

Roche

storage temp.

room temp

Related Categories

General description

Chromozym® PK is a non-radioactive amidolytic chromogenic substrate mostly used in kinetic analysis of plasma kallikrein.

Application

Use Chromozym® PK as a substrate for the determination of serine proteases, specifically plasma kallikrein in citrated plasma.

Quality

Contaminant: <0.5% free 4-nitraniline

Sequence

Benzoyl-prolyl-phenalanyl-arginine-4-nitranilide-acetate

Principle

Chromozym PK is cleaved by plasma kallikrein into a residual peptide and free 4-nitraniline, which is measured at 405 nm. The absorbance difference per minute is used for the determination of the kallikrein activity in U/ml.

Preparation Note

Activator: Mg2+ (Km = 3.0 mM, pH 7.4, 25 °C); other divalent metal ions (Mn2+, Fe2+, Zn2+)
Working concentration: Approximately 0.5 mM
Storage conditions (working solution): 2 to 8 °C
An aqueous solution of Chromozym PK (1.3 mM) is stable for at least 4 weeks if stored at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Chromozym is a registered trademark of Pentapharm AG, Basel

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Ito et al.
Clinical chemistry, 27(4), 586-593 (1981-04-01)
The amidolytic activity of activated kallikrein in plasma can be measured by use of the chromogenic substrate, S-2302 (H-D-Pro-Phe-Arg-pNA). Plasma prekallikrein was activated to kallikrein by exposure to 50 mg/L dextran sulfate in acetone/water (35/65 by vol) at 0 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service