Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10102105001

Roche

Adenosine Deaminase (ADA)

from calf intestine

Synonym(s):

ADA

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

bovine (calf) intestine

Quality Level

form

solution

specific activity

~200 units/mg protein (At 25 °C with adenosine as the substrate.)

mol wt

230-440 kDa (high)
30-47 kDa (low )

packaging

pkg of 2 mL (10 mg)

manufacturer/tradename

Roche

optimum pH

7.0-7.4

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Adenosine Deaminase (ADA) is present majorly in the proximal small intestine and the duodenum. It is also called Adenosine aminohydrolase. It is a zinc metalloenzyme and exists as a high (230−440 kDa) and low molecular weight (30−47 kDa) isoforms.
Adenosine deaminase (ADA) is a zinc metalloenzyme.

Application

Adenosine Deaminase (ADA) has been used as a component of macrophage- serum-free media (SFM) to treat macrophages.
Adenosine deaminase has been used to pre-treat primary Kupffer cells.

Biochem/physiol Actions

Adenosine Deaminase (ADA) catalyzes the deamination of adenosine analogs to the corresponding inosine analogs. It is present in high levels in colorectal cancer and is a potential marker in tuberculous pleuriti. ADA deficiency affects T-and B-lymphocytes resulting in severe combined immunodeficiency (SCID) leading to lymphocytopenia.
Adenosine deaminase (ADA) is involved in the deamination of adenosine to inosine and 2′ deoxyadenosine to 2′-deoxyinosine.

Quality

Contaminants: <0.01% alkaline phosphatase, AMP-deaminase, guanase, and nucleoside phosphorylase each

Physical form

Solution in 50% glycerol (v/v), 10 mM potassium phosphate, pH approximately 6

Preparation Note

Storage conditions (working solution): ADA (all preparations) may be dialyzed, without loss of activity, at 2 to 8 °C against 20 mM phosphate buffer, pH 7. The dialyzed preparation is stable at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Adenosine deaminase deficiency: a review
Flinn AM and Gennery AR
Orphanet Journal of Rare Diseases, 13(1), 65-65 (2018)
Pdx-1 is required for activation in vivo from a duodenum-specific enhancer
Dusing MR, et al.
The Journal of biological chemistry, 276(17), 14434-14442 (2001)
Purification and characterization of intestinal adenosine deaminase from mice
Singh L and Sharma R
Molecular and Cellular Biochemistry, 204(1-2), 127-134 (2000)
Regulation of MMP-9 expression by the A2b adenosine receptor and its dependency on TNF-alpha signaling
Chen H, et al.
Experimental Hematology, 39, 525-530 (2011)
Development of adenosine deaminase-specific IgY antibodies: Diagnostic and inhibitory application
Lupicka-Slowik A, et al.
Applied Biochemistry and Biotechnology, 184(4), 1358-1374 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service