Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

XX1014703

Millipore

Millipore Spring Clamp for Vacuum Filtration

47 mm, Aluminum

Synonym(s):

Clamp for Vacuum, Millipore Filtration Clamp, Spring Clamp for Filtration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

anodized aluminum

technique(s)

vacuum filtration: suitable

diam.

47 mm

application(s)

filtration

shipped in

ambient

General description

Millipore Glass Vacuum Filtration Accessory Product
Spring Clamp, 47 mm, Aluminum

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits and Millipore MilliSolve Kit.

Application

47 mm, Aluminum

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service