Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WX0008

Supelco

Water

suitable for HPLC, suitable for EPA 200.8 and EPA 200.7

Synonym(s):

high purity solvents

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
50202301
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
100 °C (lit.)
vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Water, HPLC

grade

HPLC grade

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

vapor pressure

23 hPa ( 20 °C)

form

liquid

shelf life

12 mo. (from manufacture date)

technique(s)

HPLC: suitable

evapn. residue

≤2 ppm

color

APHA: ≤10

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

UV absorption

λ: 200 nm Amax: ≤0.010
λ: 210 nm Amax: ≤0.010
λ: 254 nm Amax: ≤0.005
λ: 300-400 nm Amax: ≤0.005

application(s)

general analytical

shipped in

ambient

storage temp.

no temp limit

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Expiration Dated
HPLC

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Expiration dateExp 12 months from mfg dateAppearanceClear liquid, free from particulatesColor (APHA)10 maxEvaporation residue2ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)150ppt maxGradient at 210 nm0.002AU maxGradient at 254 nm0.0005AU maxOdorTo pass testRefractive index (n 25/D)1.3320-1.3340UV Abs. at 200 nm0.010AU maxUV Abs. at 210 nm0.010AU maxUV Abs. at 254 nm0.005AU maxUV Abs. at 300 - 400 nm0.005AU max

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service