Skip to Content
MilliporeSigma

TX1568

Triton X-100

Non-ionic detergent and emulsifier

Synonym(s):

Triton X-100, Octylphenol Decaethylene Glycol Ether, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
UNSPSC Code:
12161902
MDL number:
Beilstein/REAXYS Number:
2315025
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Triton X-100 Surfactant,

form

viscous liquid

Quality Segment

mol wt

micellar avg mol wt 80,000, average mol wt 625

aggregation number

100-155

color

APHA: ≤150

CMC

0.2-0.9 mM (20-25°C)

mp

6 °C

transition temp

flash point 251 °C, cloud point 65 °C, pour point ~7 °C

HLB

13.5

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Application


  • Identification of protein components of the transformation system in the cell line of immortalized human keratinocytes HaCaT exposed to surfactants.: This study utilizes Triton X-100 Surfactant to identify protein components in immortalized human keratinocytes. The research highlights the surfactant′s role in cell transformation systems and its impact on protein composition, providing insights into cellular response mechanisms. (Shkrigunov et al., 2024).

  • In situ isolation of nuclei or nuclear proteins from adherent cells: a simple, effective method with less cytoplasmic contamination.: This study presents a method using Triton X-100 Surfactant for isolating nuclei or nuclear proteins from adherent cells. The findings demonstrate reduced cytoplasmic contamination, enhancing the accuracy of nuclear protein studies. (Qin et al., 2023).

  • Inter-Regional Proteomic Profiling of the Human Brain Using an Optimized Protein Extraction Method from Formalin-Fixed Tissue to Identify Signaling Pathways.: This research employs Triton X-100 Surfactant in an optimized protein extraction method for proteomic profiling of the human brain. The study identifies critical signaling pathways, underscoring the surfactant′s utility in neuroscience research. (Davidson et al., 2023).


Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Acidity (as Na2O)%Alkalinity (as Na2O)%Cloud point (1%, water)63-69°CColor (APHA)150 maxFormClear, viscous liquidSpecific Gravity at 25/25°C1.063-1.067pH of a 1% aqueous solution5-8

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow


Still not finding the right product?

Explore all of our products under Triton X-100 Surfactant


signalword

Danger

Storage Class

10 - Combustible liquids

flash_point_f

483.8 °F - closed cup

flash_point_c

251 °C - closed cup

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - ED ENV 1 - Eye Dam. 1 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library