Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX1320R

Supelco

Spill Kit, Caustic Replacement Kit

SpillSolv®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905

pH

2.2 ( in H2O, as aqueous solution)

shipped in

ambient

storage temp.

room temp

Application

SpillSolv

Legal Information

SPILLSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service