MilliporeSigma
All Photos(4)
Key Documents
SQPAKT0CM1
Milli-Q
SQPAK TOC Quanta Cartridge
greener alternative
Produces ultrapure (Type 1) water with TOC ≤ 5 ppb. Use with the ultrapure dispensing module of Milli-Q® SQ 2Series systems.
About This Item
UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NB.35
Quality Level
packaging
pkg of 1 unit
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
compatibility
for use with Milli-Q® SQ 2Series
greener alternative category
General description
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The SQPAK TOC Quanta cartridge produces ultrapure (Type 1) water at 18.2 MΩ·cm resistivity and TOC ≤ 5 ppb.
The cartridge contains IQnano ion-exchange media to remove ions down to trace levels and high-grade synthetic activated carbon that targets traces of organic contaminants.
For use on the ultrapure dispensing module of Milli-Q® SQ 2Series configurations, including Milli-Q® SQ 200/200P and SQ 240C/240CV systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Features and Benefits
  • Easy replacement thanks to the Twist & Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
  • Ultrapure label and blue color of the SQPAK Quanta cartridge are repeated on the production station interface, to inform about its water purification purpose and guide replacement  
Legal Information
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Protocols
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
Related Content
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.