Skip to Content
MilliporeSigma

SLM-220

EmbryoMax® DMEM (1X), liquid, With 4,500mg/L Glucose, 2.25g/L Sodium Bicarb, without L-Glut and Sodium Pyruvate

The EmbryoMax DMEM with 4,500mg/L Glucose, 2.25g/L Sodium Bicarb, without L-Glut & Sodium Pyruvate is available in a 500 mL format and may be used for routine mouse embryonic stem cell culture.

Synonym(s):

Low Bicarb Formulation

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161702
NACRES:
NA.75
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells
sample type cardiac stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)

Application

EmbryoMax® DMEM (1X), liquid, with 4,500mg/L glucose, 2.25g/L sodium bicarb, without L-glut and sodium pyruvate has been used:
  • for routine mouse embryonic stem cell culture applications
  • to harbor the mouse embryonic feeder layer for mouse embryonic stem (ES) cell cultivation
  • in the preparation of chick dorsal root ganglion (DRG)-conditioned medium (CM)

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library