Skip to Content
MilliporeSigma

SLM-202

Dulbecco′s Modified Eagle′s Media 2X, With 4,500 mg/L Glucose and L-Glutamine, without NaHCO3 or Sodium Pyruvate

Synonym(s):

DMEM 2X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

The Dulbecco′s Modified Eagle′s Media 2X, With 4,500 mg/L Glucose & L-Glutamine, without NaHCO3 or Sodium Pyruvate is available in a 500 mL format & has been optimized & validated for cell culture.

Quality Segment

form

liquid

manufacturer/tradename

Specialty Media

technique(s)

cell culture | mammalian: suitable

input

sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type induced pluripotent stem cell(s)
sample type neural stem cell(s)
sample type hematopoietic stem cell(s)
sample type pancreatic stem cell(s)
sample type: human embryonic stem cell(s)
sample type cardiac stem cell(s)
sample type mesenchymal stem cell(s)

General description

The Dulbecco′s Modified Eagle′s Media 2X, With 4,500 mg/L Glucose & L-Glutamine, without NaHCO3 or Sodium Pyruvate is available in a 500 mL format. It is optimized and validated for use in cell culture.

Application

Dulbecco′s Modified Eagle′s Media 2X, With 4,500 mg/L Glucose and L-Glutamine, without NaHCO3 or Sodium Pyruvate has been used:
  • in a well plate to mix transformed or parental infrared fluorescent protein (iRFP) 3T3 fibroblasts for soft agar assay
  • to prepare collagen solution for collagen gel contraction
  • in the preparation of 5% methyl cellulose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library