Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCT151

Sigma-Aldrich

Firefly Luciferase Assay (Lyophilized)

Lyophilized flash-type luciferase assay designed for simple and efficient quantitation of firefly luciferase reporter enzyme activity from cultured cells with high sensitivity and linearity.

Synonym(s):

Firefly Luciferase Assay, Bioluminescent Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

technique(s)

cell based assay: suitable

detection method

chemiluminescent

shipped in

ambient

General description

Firefly luciferase is widely used as a reporter for studying gene regulation and function, and for pharmaceutical screening. It is a very sensitive genetic reporter due to the absence of endogenous luciferase activity in mammalian cells or tissues. Firefly luciferase is a 62,000 Dalton protein, which is active as a monomer and does not require subsequent processing for its activity. The enzyme catalyzes ATP-dependent D-luciferin oxidation to oxyluciferin, producing light emission centered at 560 nm. Light emitted from the reaction is directly proportional to the number of luciferase enzyme molecules.

The Firefly Luciferase Assay (Lyophilized) is designed for simple and efficient quantitation of firefly luciferase reporter enzyme activity from cultured cells with high sensitivity and linearity. The Firefly Assay buffer is packaged as a convenient lyophilized powder, which can be shipped at ambient temperature and stored at –20 °C instead of –80 °C. This is a flash-type luminescence assay with signal half-life of about 12 minutes.

Application

Firefly Luciferase Assay
Lyophilized flash-type luciferase assay designed for simple and efficient quantitation of firefly luciferase reporter enzyme activity from cultured cells with high sensitivity and linearity.
Research Category
Cell Signaling
Research Sub Category
Live Cell Dye

Components

1) 5X Firefly Luciferase Lysis Buffer (CS224525): 2 X 15 mL

2) Firefly Luciferase Assay Buffer (Lyophilized) (CS224526): 1000 assays

3) D-Luciferin (CS224519): 2 X 10 mg

Physical form

Liquid

Storage and Stability

Store Firefly Luciferase Assay (Lyophilized) at -20°C. The kit is stable at -20°C for at least six months from date of receipt. After reconstitution, aliquot assay buffer if necessary to avoid repeated freeze-thaw cycles; reconstituted assay buffer is stable at -20°C for at least 3 months or -80°C for at least 6 months. Firefly luciferase working solution (assay buffer + D-luciferin) should be prepared fresh on the day of assay.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service