Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCT079

Sigma-Aldrich

BioTracker CoxFluor Live Cell Dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

Assay

≥95% (HPLC)

form

lyophilized

mol wt

calculated mol wt 501.66

packaging

pkg of 1 mg

manufacturer/tradename

Millipore

storage condition

desiccated
protect from light

technique(s)

cell based assay: suitable

color

off-white

fluorescence

λex 540 nm; λem 590 nm

detection method

fluorometric

shipped in

ambient

storage temp.

−20°C

General description

Cyclooxygenase (COX), also known as Prostaglandin-endoperoxide synthase (PTGS), is the key enzyme in prostaglandin biosynthesis. There are two isozymes of cyclooxygenase: a constitutive COX-1 and an inducible COX-2, which differ in their regulation of expression and tissue distribution. COX-2 overexpression is prominent in inflammatory diseases, neurodegenerative disorders, and cancer. Directly monitoring COX-2 activity within its native environment poses an exciting approach to investigate the effect of the local environments on protein activity.

BioTracker CoxFluor Live Cell Dye is an isoform selective activity-based sensor for COX-2. The probe may be used for COX-2 inhibitor screening assays and for monitoring COX-2 activity in live cells by confocal microscopy or flow cytometry. CoxFluor strategically links a natural substrate with a dye precursor to engage both the cyclooxygenase and peroxidase activities of COX-2. This catalyzes the release of resorufin and the natural product, as supported by molecular dynamics and ensemble docking. CoxFluor enabled the detection of oxygen dependent changes in COX-2 activity that are independent of protein expression within live macrophage cells.

Spectral Properties
Absorbance: 540 nm
Emission: 590 nm

Reference:
1. Yadav, A. K., Reinhardt, C. J., Arango, A. S., Huff, H. C., Dong, L., Malkowski, M. G., ... & Chan, J. (2020). An Activity?Based Sensing Approach for the Detection of Cyclooxygenase?2 in Live Cells. Angewandte Chemie, 132(8), 3333-3340.

Application

Purity ≥ 95% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

Fluorescent probe for monitoring COX-2 activity in live cells by confocal microscopy or flow cytometry.

Physical form

Lyophilized. Off-white solid.

Storage and Stability

Store at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

Live cell fluorescent imaging

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anuj K Yadav et al.
Angewandte Chemie (International ed. in English), 59(8), 3307-3314 (2019-12-20)
Cyclooxygenase-2 (COX-2) overexpression is prominent in inflammatory diseases, neurodegenerative disorders, and cancer. Directly monitoring COX-2 activity within its native environment poses an exciting approach to account for and illuminate the effect of the local environments on protein activity. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service