Skip to Content
MilliporeSigma

SCT035

BioTracker 515 Green ONOO- Dye

Live cell imaging dye for peroxynitrite (ONOO-) used to detect reactive oxygen species (ROS) generated from the interaction of nitric oxide (NO) and superoxide (O).

Synonym(s):

Live cell imaging probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

-10 to -25°C

General description

Peroxynitrite (ONOO-) is a strong oxidant generated from the interaction of nitric oxide (NO) and superoxide (O). The in vivo formation of this compound has recently been demonstrated in macrophages, neutrophils and endothelial cells. The role of ONOO- in inflammatory diseases is further supported by the detection of immunoreactive nitrotyrosine on proteins at sites of inflammation.

The BioTracker 515 Green ONOO- Dye is a fluorescent probe that detects peroxynitrite (ONOO-) in living cells. Fluorescent intensity is not increased by the existence of hydroxyl radical, singlet oxygen, hydrogen peroxide, hypochlorite, nitric oxide or superoxide.

Spectral Properties
Absorbance: 490nm
Emission: 515nm

Preparation Note

Store BioTracker 515 Green ONOO- Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library