Skip to Content

SCT021

BioTracker 609 Red Ca2+ AM Dye

Live cell imaging calcium (Ca2+) indicator for neuroscience and cancer applications.

Synonym(s):

Live cell calcium detection probe, Live cell imaging probe, Red fluorescent calcium indicator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

storage temp.

-10 to -25°C

General description

Calcium (Ca2+) play a vital role in the physiology and biochemistry of the cell. Calcium ions play an important role in signal transduction pathways, neurotransmitter release from neurons, contraction of all muscle cell types, and in fertilization. Extracellular calcium is also important for maintaining the potential difference across excitable cell membranes, as well as proper bone formation.

The BioTracker 609 Red Ca2+ AM Dye is a live cell red fluorescent calcium indicator. Fluorescence intensity of this fluorescent probe reversibly changes depending on Ca2+ concentration. It has red fluorescence with maximum emission at 609 nm (Excitation 590nm) and is suitable for detection of micromolar- to submicromolar-range calcium ions. The dye is hydrolyzed by intracellular esterases to become soluble which equally distributes in the cytoplasm. This feature enables detection of intracellular calcium concentration changes with superior accuracy compared to other calcium indicators.

Spectral Properties
Absorbance: 595nm
Emission: 609nm

Preparation Note

Store BioTracker 609 Red Ca2+ AM Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

1202.4 °F

flash_point_c

650.2 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library