Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCR104

Sigma-Aldrich

Accutase®XL Lyophilized Accutase

AccutaseXL is a lyophilized form of Accutase with extended stability used for the routine detachment of cells from standard tissue culture plasticware

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160801
NACRES:
NA.78

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

General description

Accutase is a cell detachment solution of proteolytic and collagenolytic enzymes that is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including SmartPlastic. Accutase is direct replacement for trypsin cell detachment solution and does not require enzyme inactivation with serum. Accutase does not contain mammalian or bacterial derived products.

AccutaseXL is a lyophilized form of Accutase with extended stability. AccutaseXL, once reconstituted, will make 500ml of complete 1X Accutase solution. Note: read detailed reconstitution protocol found in the product datasheet before using this product.
Product Source: Accutase is derived from a non-mammalian (invertebrate) source.

Application

Accutase has been shown to be effective on a wide variety of cell types including: fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, vascular smooth muscle cells, hepatocyte progenitors, primary chick embryo neuronal cells, bone marrow stem cells, adherent CHO and BHK cells, macrophages, 293 cells, L929 cells, immortalized mouse testicular germ cells, 3T3, Vero, COS, HeLa, NT2, MG63, M24 and A375 metastatic melanoma, gliomas U251 and D54, HT1080 fibrosarcoma cells, and Sf9 insect cells.

Quality

pH: 6.8-7.8
Activity Assay: 500-20 units/ml (1 unit will release 1 µmole pNA from SUCR-pNA per minute at 37˚C)
Functional Assay: Cell detachment assay

Physical form

1X Accutase enzymes in Dulbecco’s PBS (0.2 g/L KCl, 0.2 g/L KH2PO4, 8 g/L NaCl, and 1.15 g/L Na2HPO4) containing 0.5 mM EDTA4Na and 3 mg/L Phenol Red.

Storage and Stability

Store lyophilized AccutaseXL at -20°C for up to 4 months from date of receipt.
Once reconstituted in water, liquid Accutase can be aliquoted and frozen at -20°C for 2 years or stored for 2 months at 4°C. It is recommended to thaw liquid Accutase at 4°C overnight or in a bath of cool water.

Legal Information

Accutase is a registered trademark of Innovative Cell Technologies, Inc.
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service