Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCM105

Sigma-Aldrich

3dGRO® L-WRN Conditioned Media Supplement

Synonym(s):

Conditioned Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

manufacturer/tradename

Millipore

technique(s)

cell based assay: suitable

color

clear red

shipped in

ambient

storage temp.

−20°C

Application

Appearance (color): clear, red liquid
Endotoxin: <2 EU/mL
Sterility Tested: No Growth/Pass
Media are tested for function in supporting growth of human intestinal organoids
Media are tested for levels of growth factors by ELISA
The 3dGRO® L-WRN Conditioned Media Supplement is a formulation based on DMEM/F-12 PLUS medium containing the 3 growth factors Wnt3a, R-spondin-3, and Noggin. These factors act upon the Wnt signaling pathway, which plays a critical role in sustaining the proliferation of LGR5+ crypt stem cells within organoids. Wnt activation is thus essential for in vitro growth of human gut organoids. L-WRN conditioned medium is widely used as a component of the culture medium for supporting the growth of human intestinal organoids including the 3dGRO® gastrointestinal organoid biobank (SCC310-SCC367). Applications for L-WRN conditioned medium include but are not limited to the expansion of human intestinal gut organoids and other epithelial organoid cell types.



Source


The 3dGRO® L-WRN Conditioned Media Supplement is derived from the Mus musculus L-M(TK-) cell line secreting mouse Wnt3a, R-spondin-3 and Noggin. L-WRN Conditioned Medium may contain trace amounts of mouse DNA.

Features and Benefits

L-WRN conditioned media supplement containing the 3 growth factors Wnt3a (W), R-spondin-3 (R), and Noggin (N) for organoid cultures.

Storage and Stability

The 3dGRO® L-WRN Conditioned Media Supplement should be stored at -20°C. When ready to use, thaw the L-WRN Conditioned Medium overnight at 2-8°C in the dark. Thawed medium is good for two weeks when stored at 2-8°C. If media will not be used within two weeks, aliquot unused media and store at -20°C. After thawing refrozen aliquots, do not re-freeze media.

Legal Information

3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.

Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.

Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.

Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.

Related Content

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service