Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
manufacturer/tradename
Chemicon®
technique(s)
cell culture | stem cell: suitable
input
sample type mesenchymal stem cell(s)
shipped in
liquid nitrogen
storage temp.
-140 to -196°C
General description
Application
EMD Millipore’s Human Mesenchymal Stem Cells (MSC) are derived from MEL-1 human embryonic stem cells (hESC). The cells proliferate as an adherent cell monolayer and can be expanded for up to 10 passages. Human ESC-derived MSC express the appropriate MSC markers including H-CAM (CD44), STRO-1, Thy-1 (CD90), and M-CAM (CD146) and do not express hematopoietic cell surface markers, CD14 (present on leukocytes) and CD19 (present on B-lymphocytes) and pluripotent markers, Oct-4 and SSEA-4. Cells can readily differentiate into adipocytes and osteoctyes.
EMD Millipore’s cryopreserved Human Mesenchymal Stem Cells derived from human ESC (Cat. No. SCC036) can be optimally expanded in low-serum media (FibroGRO LS Complete Media Kit for Culture of Human Fibroblasts, Cat. No. SCMF002). For serum-free applications, cells can also be expanded in serum-free media (FibroGRO Complete Media Kit for Culture of Human Fibroblasts, Cat. No. SCMF001).
Stem Cell Research
Preparation Note
M
Analysis Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Includes 1 million viable cells of Human Mesenchymal Stem Cells (derived from hES cells). | description These primary bone marrow derived Rat Mesenchymal Stem Cells isolated from the iliac crest of adult Fisher 344 rats provide a convenient source of multipotent rat mesenchymal stem cells. | description Human mesenchymal stem cells derived from bone marrow that have a capacity for expansion in vitro while maintaining their potential for differentiation to multiple lineages including adipocytes, osteoblasts and chondrocytes. | description The PluriStem 129S6 Murine ES cell line is derived from the widely used 129/S6/SvEv strain of mice (at 3.5 days p.c.) & is intended for injection into black or albino host blastocysts. |
| technique(s) cell culture | stem cell: suitable | technique(s) cell culture | stem cell: suitable | technique(s) cell culture | mammalian: suitable, cell culture | stem cell: suitable, cell differentiation: suitable | technique(s) cell culture | stem cell: suitable |
| shipped in liquid nitrogen | shipped in liquid nitrogen | shipped in liquid nitrogen | shipped in liquid nitrogen |
| manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® | manufacturer/tradename - | manufacturer/tradename Chemicon®, PluriSTEM® |
| storage temp. -140 to -196°C | storage temp. -140 to -196°C | storage temp. -140 to -196°C | storage temp. -140 to -196°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| input sample type mesenchymal stem cell(s) | input sample type mesenchymal stem cell(s) | input sample type mesenchymal stem cell(s) | input sample type: mouse embryonic stem cell(s) |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


