Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCC007

Sigma-Aldrich

ReNcell CX Human Neural Progenitor Cell Line

ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons & glial cells.

Synonym(s):

Human NSC Line, Human Neural Stem Cell Line, Human Neural Stem Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.75

biological source

human

Quality Level

manufacturer/tradename

Chemicon®
ReNcell

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

liquid nitrogen

General description

Product Source: Human cortical brain tissue
ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells. ReNcell CX was derived from the cortical region of human fetal brain. Immortalized by retroviral transduction with the c-myc oncogene, this cell line grows rapidly as a monolayer on laminin with a doubling time of 20-30 hours. Karyotype analyses indicate that the ReNcell CX retains a normal diploid karyotype in culture even after prolonged passage (>45 passages). ReNcell CX was developed by the ReNeuron Group plc, a biotech company that specializes in using human somatic stem cells for therapeutics. ReNcell CX may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ReNcell CX cells have been obtained in a legal and ethical manner, compliant with current local informed consent procedures.

ReNcell Now Used in a 3D Cell Model of Alzheimer′s Disease!
Tanzi, R and Kim, DY et. al. A three-dimensional human neural cell culture model of Alzheimer′s disease. Nature. 2014 Nov 13;515(7526):274-8.
(Published in Nature)

Cell Line Description

Neural Stem Cells

Application

Research Category
Stem Cell Research

Components

> 1x106 viable ReNcell CX cells

Quality

Each lot of ReNcell CX cells has been validated for high level of expression of Nestin and Sox 2 and for their self-renewal and multi-lineage differentiation capacities (please refer to datasheet figures). Cells also display normal karyotype as assessed by chromosome spread and tested negative for mycoplasma. Maternal blood has been screened for HIV, HTLV, Hepatitis B and C.

Storage and Stability

When stored at the recommended storage conditions (liquid nitrogen), ReNcell CX cells are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date. We recommend that the cells be used within ten passages.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the "Academic Use Agreement" as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Luo et al.
Cells, 10(3) (2021-03-07)
Human adipose-derived stromal cells (ADSCs) are receiving unprecedented attention as a potential cellular source for regenerative medicine-based therapies against various diseases and conditions. However, there still have significant issues concerning the translational development of ADSC-based therapies, such as its heterogeneity
Greg Maguire et al.
Physiological reports, 7(9), e14072-e14072 (2019-05-03)
Evidence suggests that adult stem cell types and progenitor cells act collectively in a given tissue to maintain and heal organs, such as muscle, through a release of a multitude of molecules packaged into exosomes from the different cell types.
Qi Gu et al.
Advanced healthcare materials, 5(12), 1429-1438 (2016-03-31)
Direct-write printing of stem cells within biomaterials presents an opportunity to engineer tissue for in vitro modeling and regenerative medicine. Here, a first example of constructing neural tissue by printing human neural stem cells that are differentiated in situ to
Sebastian Peters et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 18(3), 1963-1979 (2021-04-17)
Adult neurogenesis is a target for brain rejuvenation as well as regeneration in aging and disease. Numerous approaches showed efficacy to elevate neurogenesis in rodents, yet translation into therapies has not been achieved. Here, we introduce a novel human TGFβ-RII
Suojun Zhang et al.
International journal of oncology, 45(5), 1989-1996 (2014-09-02)
Glioma stem cells (GSCs), characterized by self-renewal, multi-potentiality and tumorigenicity, are responsible for the tumor propagation, recurrence and resistance to traditional treatments, representing a critical therapeutic target. Neural stem cells (NSCs) possess inherent tropism to brain tumor cells and inhibit

Articles

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

See All

Protocols

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service