Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SANKIT003

Milli-Q

Sanitization Kit

For RiOs / Elix® Essential water purification systems

Synonym(s):

Sanitization Kit for RiOs / Elix® Essential water purification systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14
Pricing and availability is not currently available.

feature

3 Sanitization steps (duration)
65 min for cycle 1
20 min for cycle 2
vary with tank size/STARTPAK

packaging

pkg of 1 kit

fitting

6 mm inlet connection
6 mm outlet connection

compatibility

for use with Elix® Essential UV
for use with Elix® Essential
for use with RiOs Essential

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Sanitization Kit is a water purification consumable for chemical sanitization of water purification systems.
Sanitization kit for use with RiOs / Elix® Essential water purification systems.

Features and Benefits

Genuine consumable for the sanitization of RiOs / Elix® Essential water purification systems.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service