Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RW1914250

Millipore

MCE Membrane Filter, 1.2 μm Pore Size

Millipore, filter diam. 142 mm, hydrophilic

Synonym(s):

Reinforced Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
web cellulose membrane
white filter

Quality Level

description

142 mm diameter, mixed cellulose esters, 1.2 µm pore size, polyester web

feature

hydrophilic

manufacturer/tradename

Millipore

parameter

260 mL/min-cm2 water flow rate
70 °C max. temp.

filter diam.

142 mm

thickness

200 μm

pore size

1.2 μm pore size
75 % porosity

shipped in

ambient

General description

Filter Code: RW19
Filter Type: Screen filter
Reinforced prefilter membranes can be used to remove contaminants from a variety of liquids and gases. The filters have a high dirt-holding capacity and a low pressure drop, which makes them ideally suited for reducing contaminant levels in advance of sterilizing-grade filters. Unlike traditional prefilter materials that contain asbestos or fiberglass, our reinforced prefilter membranes are made from non-shedding materials. These filters retain contaminants on the surface of the cellulose membrane.

Features & Benefits:
•Prefilter with high dirt-loading capacity for greater throughput
•Non-shedding substrate will not unload fibers downstream

Applications:
Prefiltration Ahead of Sterilizing-Grade Filters

Application

Prefiltration before 1.2 μm membrane filtration

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service