Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX0996

Supelco

o-Phosphoric Acid

85% HPLC, Meets ACS Specifications

Synonym(s):

Phosphoric acid, Orthophosphoric Acid, Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352301

Product Name

o-Phosphoric Acid 85% HPLC, Meets ACS Specifications

grade

ACS reagent

Quality Level

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

form

viscous liquid

concentration

85% phosphoric acid

impurities

≤0.001% Heavy metals (as Pb)
≤0.001% Insoluble matter
≤0.001% Sulfate (SO4)
≤0.001% Volatile acids (as acetic acid)
≤0.002% Antimony (Sb)
≤0.002% Calcium (Ca)
≤0.002% Magnesium (Mg)
≤0.003% Iron (Fe)
≤0.005% Potassium (K)
≤0.025% Sodium (Na)
≤0.5 ppm Arsenic (As)
≤0.5 ppm Manganese (Mn)
≤3 ppm Chloride (Cl)
≤5 ppm Nitrate (NO3)

color

APHA: ≤10

pH

<0.5 (20 °C, 100 g/L in H2O)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

UV absorption

λ: 220 nm Amax: ≤0.05
λ: 254 nm Amax: ≤0.04
λ: 300 nm Amax: ≤0.02

shipped in

ambient

storage temp.

room temp

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Meets ACS Specifications
PX0996-6 is packaged in poly coated glass bottles

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Analysis Note

Antimony (Sb)0.002% maxArsenic (As)0.5ppm maxAssay85.0% minCalcium (Ca)0.002% maxChloride (Cl)3ppm maxColor (APHA)10 maxFormClear syrupy liquidHeavy metals (as Pb)0.001% maxInsoluble matter0.001% maxIron (Fe)0.003% maxMagnesium (Mg)0.002% maxManganese (Mn)0.5ppm maxNitrate (NO3)5ppm maxPhosphorous or hypophosphorous acidTo pass testPotassium (K)0.005% maxReducing substancesTo pass testSodium (Na)0.025% maxSulfate (SO4)0.001% maxUV Absorbance at 220 nm (1M, 1cm, water)0.05AU maxUV Absorbance at 254 nm (1M, 1cm, water)0.04AU maxUV Absorbance at 300 nm (1M, 1cm, water)0.02AU maxVolatile acids (as acetic acid)0.001% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service