Skip to Content
MilliporeSigma
All Photos(1)

Documents

PX0427

Supelco

Petroleum Ether BP 65-100°C

Reagent

Synonym(s):

Petroleum ether, Ligroine,Petroleum Spirit, Ligroine, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
EC Index Number:
232-453-7

Quality Level

form

liquid

evapn. residue

≤0.001%

color

APHA: ≤10

bp

60-90 °C

transition temp

flash point -7 °C

density

0.73 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Reagent

Analysis Note

AcidTo pass testAppearanceTo pass testBoiling range60-90°CColor (APHA)10 maxEvaporation residue0.001% maxHeavy oils and fatsTo pass testOdorTo pass testSulfur compounds (Doctor test)Negative

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service