Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PRCYTOMAG-40K

Millipore

MILLIPLEX® Non-Human Primate Cytokine Magnetic Bead Panel - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.

Synonym(s):

Luminex® non-human primate cytokine immunoassay, Millipore non-human primate cytokine panel, Monkey cytokine multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

nonhuman primates

manufacturer/tradename

Milliplex®

assay range

accuracy: 70-101%
standard curve range: 12.2-50,000 pg/mL
(IL-10, IL-18)

standard curve range: 2.4-10,000 pg/mL
standard curve range: 4.9-20,000 pg/mL
(IL-4)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

ambient

General description

The immune system is an organized complex network of biological structures and processes that protect against disease. For example, cytokine and chemokines mediate interactions between cells directly, regulating target immune cell responses. These responses result in inflammation where the immune system attempts to eradicate foreign antigens and begin the healing process. Consequently, the inflammatory process plays a key protective role in immunity. In addition, cytokine and chemokine research is essential in understanding the immune system and its multi-faceted response to most antigens, as well as disease states such as autoimmune disease, allergic reactions, sepsis, and cancer.

The MILLIPLEX® Non-Human Primate Cytokine panel is to be used for the simultaneous quantification of any or all of the following in tissue/cell lysate and culture supernatant samples and serum or plasma samples: G-CSF, GM-CSF, IFNγ, IL-1β, IL-1ra, IL-2, IL-4, IL-5, IL-6, IL8, IL-10, IL-12/23(p40), IL-13, IL-15, IL-17A, MCP-1, MIP-1β, MIP-1α, sD40L, TGF-α, TNF-α, VEGF, and IL-18.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
The multiplexed assay panel has been tested with samples from multiple non-human
primate species, please see protocol for details.
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: G-CSF, GM-CSF, IFN-γ, IL-1RA, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-8, IL-10, IL-12/23 (p40), IL-13, IL-15, IL-17, IL-18, MCP-1, MIP-1α, MIP-1β, sCD40L, TGF-α, TNF-α, VEGF
  • Recommended Sample type: Serum, plasma, and cell culture
  • Recommended Sample dilution: Neat
  • Assay Run Time: Overnight or one day. For best results, an overnight incubation is recommended
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities for individual cytokines/chemokines
Standard Curve Range: Refer to QC analysis sheet for exact concentration (sCD40L, TNFα)

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

G-CSF, GM-CSF, IFN-γ, IL-1ra, IL-1ß, IL-2, IL-5, IL-6, IL-8, IL-12/23 (p40), IL-13, IL-15, IL-17, MCP-1, MIP-1α, MIP-1ß, TGF-α, VEGF
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Verreault et al.
Antimicrobial agents and chemotherapy, 63(5) (2019-02-21)
Staphylococcal enterotoxin B (SEB) is a protein exotoxin found on the cell surface of Staphylococcus aureus that is the source for multiple pathologies in humans. When purified and concentrated in aerosol form, SEB can cause an acute and often fatal
Tetsu Oura et al.
Cell transplantation, 25(7), 1331-1341 (2015-09-05)
We have previously reported successful induction of transient mixed chimerism and long-term acceptance of renal allografts in MHC mismatched nonhuman primates. In this study, we attempted to extend this tolerance induction approach to islet allografts. A total of eight recipients
Johannes Lutz et al.
NPJ vaccines, 2, 29-29 (2017-12-22)
mRNA represents a promising new vaccine technology platform with high flexibility in regard to development and production. Here, we demonstrate that vaccines based on sequence optimized, chemically unmodified mRNA formulated in optimized lipid nanoparticles (LNPs) are highly immunogenic and well
Whitney A Nichols et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 74(6), 508-522 (2015-08-21)
HIV infection among women is frequently modeled in female rhesus macaques. Longitudinal studies on genital compartment and hormonal factors that can influence susceptibility to SIV infection are lacking in this animal model. Genital specimens and menstruation of indoor-housed female rhesus
Staci Sabnis et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(6), 1509-1519 (2018-04-15)
The success of mRNA-based therapies depends on the availability of a safe and efficient delivery vehicle. Lipid nanoparticles have been identified as a viable option. However, there are concerns whether an acceptable tolerability profile for chronic dosing can be achieved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service