Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
5
biological source
rat
Quality Level
antibody form
purified immunoglobulin
clone
polyclonal
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
shipped in
wet ice
target post-translational modification
unmodified
General description
Source: Normal rat serum
Physical form
Format: Purified
Sterile liquid. Buffer = 0. 01M Sodium Phosphate, 0.25M NaCl, pH 7.6.
Analysis Note
Precipitin line against goat anti-rat whole serum is the same as that against goat anti rat IgG, Fc fragment specific when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hiroaki Sato et al.
The Journal of dermatology, 31(3), 200-217 (2004-06-10)
Tacalcitol (1,24(R)(OH)2D3, TV-02) inhibited the TPA-induced inflammatory cell infiltration (largely neutrophils) histopathologically and myeloperoxidase (MPO) activity dose-dependently. Tacalcitol inhibited the mRNA expression and protein production of TPA-induced macrophage inflammatory protein-2 (MIP-2) and KC, the functional analogue of human interleukin (IL)-8
Isolation of nuclei from label-retaining cells and measurement of their turnover rates in rat colon.
Kim, SJ; Cheung, S; Hellerstein, MK
American Journal of Physiology. Cell Physiology null
FGFR3 contributes to intestinal crypt cell growth arrest.
Sandrine Arnaud-Dabernat,Deepak Yadav,Nora Sarvetnick
Journal of Cellular Physiology null
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PP68 | 04053252505515 |