Skip to Content
MilliporeSigma

Skip To

PLTK06210

Regenerated Cellulose Membrane Filter, 30 kDa NMWCO

Ultracel®, filter diam. 63.5 mm

Synonym(s):

Ultrafiltration Discs, 30 kDa NMW, Ultrafiltration membrane discs, Regenerated Cellulose Membrane Filter, 30 kDa NMWCO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104903
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
30 kDa NMWCO
Material:
regenerated cellulose membrane
Feature:
hydrophilic
Diam.:
63.5 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ultracel® 30 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 63.5 mm Amicon® Bioseparations

material

regenerated cellulose membrane

Quality Segment

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting), protein purification: suitable (concentration)

diam.

63.5 mm

filter diam.

63.5 mm

filtration area

28.7 cm2

matrix

Ultracel®

pore size

30 kDa NMWCO

shipped in

ambient

General description

Ultrafiltration membranes are typically made from regenerated cellulose or polyethersulfone (PES). As a rule, the membrane nominal molecular weight cut-off (NMWCO) should be 2-3 times smaller than the molecular weight of the solute to be retained when using regenerated cellulose.

Application

Ultrafiltration Discs, 30 kDa NMW has been used with Amicon® stirred-cell concentrator for mammalian cell protein purification. It has also been used in ultrafiltration units for xylanase inhibitor fractionation.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLTK02510PLTK04310PLTK04710
description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 63.5 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 25 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 44.5 mm Amicon® Bioseparations

description

Chemistry: Regenerated Cellulose (RC) Filter Diameter: 47 mm Amicon® Bioseparations

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

manufacturer/tradename

Ultracel®

pore size

30 kDa NMWCO

pore size

30 kDa NMWCO

pore size

30 kDa NMWCO

pore size

30 kDa NMWCO

diam.

63.5 mm

diam.

25 mm

diam.

44.5 mm

diam.

47 mm

material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

material

regenerated cellulose membrane

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

-

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters