Skip to Content

PLAC15005

Regenerated Cellulose Membrane Filter, 1 kDa NMWCO

Ultracel®, filter diam. 150 mm

Synonym(s):

Ultrafiltration Discs, 1 kDa NMW, Regenerated Cellulose Membrane Filter, 1 kDa NMWCO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104903
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
1 kDa NMWCO
Material:
regenerated cellulose membrane
Feature:
hydrophilic
Diam.:
150 mm
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ultracel® 1 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 150 mm Amicon® Bioseparations

material

regenerated cellulose membrane

Quality Segment

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

Diafiltration: suitable, protein clean-up: suitable (desalting), protein purification: suitable (concentration)

diam.

150 mm

filter diam.

150 mm

filtration area

177 cm2

matrix

Ultracel®

pore size

1 kDa NMWCO

shipped in

ambient

General description

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel regenerated cellulose and Biomax polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting
Device Configuration: Ultrafiltration Discs
Filter Code: PLAC
Filter Type: Ultrafiltration

Application

1 kDa NMWL, Ultracel regenerated cellulose, 150 mm diameter

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library