Skip to Content
MilliporeSigma

P3C005C01

Millipore

Pellicon® 3 Cassette with ULTRACEL® 5 kDa Membrane, C screen, 0.11 m2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602

material

polypropylene cap (end cap)

Quality Level

product line

Pellicon®

technique(s)

buffer exchange: suitable
protein purification: suitable

H

2.3 cm (0.9 in.)

L

20.6 cm (8.1 in.)

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service