Skip to Content
MilliporeSigma

OP33

Anti-p53 (Ab-5) (Wild type) Mouse mAb (PAb1620)

liquid, clone PAb1620, Calbiochem®

Synonym(s):

Anti-p53 Antibody, Mouse Anti-p53, p53 Detection Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
PAb1620, monoclonal
Species reactivity:
human, mouse, rat, primate, bovine
Application:
Citations:
20
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb1620, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse, rat, primate, bovine

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Frozen Sections (see applications references)
Immunoblotting (not recommended)
Immunofluorescence (1-20 μg/mL)
Immunoprecipitation (1 μg per sample)
Paraffin Sections (5 μg/mL, heat pre-treatment required))

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0 Ag14 myeloma cells. Recognizes the ~53 kDa wild-type p53 protein.
Recognizes the ~53 kDa wild-type p53 protein in its native conformation in Hs27 cells and breast carcinoma tissue. Does not recognize mutant or denatured p53 protein.
This Anti-p53 (Ab-5) (Wild type) Mouse mAb (PAb1620) is validated for use in Frozen Sections, Immunoblotting, IF, IP, Paraffin Sections for the detection of p53 (Ab-5) (Wild type).

Immunogen

vLM mouse tumor cells

Application

Frozen Sections (see applications references)

Immunoblotting (not recommended)

Immunofluorescence (1-20 μg/ml)

Immunoprecipitation (1 μg per sample)

Paraffin Sections (5 μg/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Analysis Note

Positive Control
Hs27 cells or breast carcinoma tissue

Other Notes

El-Deiry, W.S., et al. 1994. Cancer Res.54, 1169.
Greenblatt, M.S., et al. 1994. Cancer Res.54, 4855.
Barak, Y., et al. 1993. EMBO J.12, 461.
Kastan, M.B., et al. 1992. Cell71, 587.
Kuerbitz, S.J. 1992. Proc. Natl. Acad. Sci. USA89, 7491.
Lane, D.P. 1992. Nature358, 15.
Kastan, M.B., et al. 1991. Cancer Res.51, 6304.
Ball, R.K., et al. 1984. EMBO J.3, 1485.
Wild-type p53 has a short half-life and is present in low amounts in cells. Increasing the amount of sample to be immunoprecipitated and applied to the gel may help for visualization. Short incubation times with 35S-Met(≤ 1 hr) will help reduce background. p53 (Ab-5) may also be used to visualize p53 in cytospins or cultured cells using standard horseradish peroxidase or immunofluorescence detection techniques. p53 (Ab-5) preferentially immunoprecipitates wild-type p53; it should not precipitate mutant or denatured p53. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Hama et al.
Journal of pharmacological sciences, 110(4), 493-496 (2009-08-05)
We have previously demonstrated an important role of influx of Cl(-) rather than Ca(2+) in acute excitotoxicity in adult rat retina. As p53 has been implicated in delayed apoptotic cell death, here we examined the appearance of p53 immunoreactivity in
Precise pancreatic cancer therapy through targeted degradation of mutant p53 protein by cerium oxide nanoparticles.
Zhang, et al.
Journal of Nanobiotechnology, 21, 117-117 (2023)
Maria L Gomez et al.
Oncogene, 38(29), 5751-5765 (2019-06-22)
We previously reported that the dismutase SOD1 is overexpressed in breast cancer. However, whether SOD1 plays an active role in tumor formation in vivo has never been demonstrated. Further, as luminal cells of normal breast epithelial cells are enriched in
Hyun-Lim Kim et al.
Molecules and cells, 38(4), 312-317 (2015-03-31)
Depletion of intracellular zinc by N,N,N',N'-tetrakis(2-pyridylmethyl) ethylenediamine (TPEN) induces p53-mediated protein synthesis-dependent apoptosis of mouse cortical neurons. Here, we examined the requirement for poly(ADP-ribose) polymerase (PARP)-1 as an upstream regulator of p53 in zinc depletion-induced neuronal apoptosis. First, we found
Alejandro Parrales et al.
Nature cell biology, 18(11), 1233-1243 (2016-10-28)
Stabilization of mutant p53 (mutp53) in tumours greatly contributes to malignant progression. However, little is known about the underlying mechanisms and therapeutic approaches to destabilize mutp53. Here, through high-throughput screening we identify statins, cholesterol-lowering drugs, as degradation inducers for conformational

Global Trade Item Number

SKUGTIN
OP33-100UG04055977208443
OP33-20UG07790788053901

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service