Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OP133

Sigma-Aldrich

Anti-IKKα Mouse mAb (14A231)

liquid, clone 14A231, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

14A231, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

monkey, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHUK(1147)

General description

Immunoaffinity purified mouse monoclonal antibody. Recognizes the ~85 kDa IKKα protein.
Recognizes the ~85 kDa IKKα protein in Daudi cells.
This Anti-IKKα Mouse mAb (14A231) is validated for use in Immunoblotting for the detection of IKKα.

Immunogen

Human
a recombinant protein consisting of full-length human IKKα/IKK1 fused to a His•Tag sequence

Application


Immunoblotting (1-2 g/ml, chemiluminescence)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.2% gelatin, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Daudi cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Zandi, E., et al. 1998 Science281, 1360.
Didonato, J.A., et al. 1997 Nature388, 548.
Lee, F.S., et al. 1997 Cell88, 213.
Mercurio, F., et al. 1997 Science278, 860.
Regnier, C.H., et al. M. 1997 Cell90, 373.
Verma, I. and Stevenson, J.K. 1997 Proc. Nat. Acad. Sci. USA94, 11758.
Verma, I.M., et al. 1995 Genes. Dev.9, 2723.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faiyaz Ahmad et al.
Scientific reports, 6, 28056-28056 (2016-06-21)
Activation of inflammation in white adipose tissue (WAT), includes infiltration/expansion of WAT macrophages, contributes pathogenesis of obesity, insulin resistance, and metabolic syndrome. The inflammasome comprises an intracellular sensor (NLR), caspase-1 and the adaptor ASC. Inflammasome activation leads to maturation of
Michael L Kamradt et al.
Cell death & disease, 12(3), 271-271 (2021-03-17)
Cancers, including glioblastoma multiforme (GBM), undergo coordinated reprogramming of metabolic pathways that control glycolysis and oxidative phosphorylation (OXPHOS) to promote tumor growth in diverse tumor microenvironments. Adaptation to limited nutrient availability in the microenvironment is associated with remodeling of mitochondrial
Cristina Aguilera et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(47), 16537-16542 (2004-11-13)
The NF-kappaB pathway plays a pivotal role in proliferation, differentiation, apoptosis, and immune responses in mammals. The NF-kappaB inhibitor, IkappaB, has classically been characterized for its ability to sequester NF-kappaB transcription factors in the cytoplasm. Nevertheless, a nuclear fraction of
Lindsay Bennett et al.
International journal of cancer, 140(7), 1633-1644 (2016-12-23)
The aim of our study was to examine the relationship between tumour IKKα expression and breast cancer recurrence and survival. Immunohistochemistry was employed in a discovery and a validation tissue microarray to assess the association of tumour IKKα expression and
Justin N Keeney et al.
Journal of immunology (Baltimore, Md. : 1950), 210(8), 1123-1133 (2023-03-08)
NF-κB-inducing kinase (NIK), which is essential for the activation of the noncanonical NF-κB pathway, regulates diverse processes in immunity, development, and disease. Although recent studies have elucidated important functions of NIK in adaptive immune cells and cancer cell metabolism, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service