Skip to Content
MilliporeSigma

Skip To

NS02L

Normal Goat Serum

This Normal Goat Serum is validated for use as a negative control in any application.

Synonym(s):

Goat serum, Normal serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352202
Biological source:
goat
Form:
lyophilized
Shipped in:
ambient
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

form

lyophilized

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(For use as a negative control in any application.)

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

General description

Normal carrier sera from healthy, non-immunized goats.

Application

For use as a negative control in any application.

Physical form

Lyophilized serum.

Preparation Note

Reconstitute the lypohilized serum in 1 ml sterile distilled H₂O with occasional gentle mixing for at least 2 h at 4°C. Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) with 0.1% azide for short-term storage. Avoid freeze/thaw cycles of solutions.

Other Notes

Normal serum is provided for use as a blocking agent in immunoassays or as negative control for any immunoassay in conjunction with polyclonal serum from Calbiochem brand. For use as a negative control, perform experiments in parallel using primary antibody or normal serum at the same concentration. A signal obtained with the normal serum indicates a nonspecific interaction; any additional signal with the primary antibody indicates a specific antibody-antigen interaction.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
566380NS01LNS03L
form

lyophilized

form

lyophilized solid

form

lyophilized

form

lyophilized

biological source

goat

biological source

goat

biological source

rabbit

biological source

mouse

shipped in

ambient

shipped in

wet ice

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters