Skip to Content
MilliporeSigma

NM0023

NovaSeal Die Kits

1/2 in. ID x 3/4 in OD

Synonym(s):

Cutting Die for Electrical Crimping Tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32010310
UNSPSC Code:
41121815
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes - NM0002PK100



Still not finding the right product?

Explore all of our products under NovaSeal Die Kits




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library