Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NE1004

Sigma-Aldrich

Anti-Apolipoprotein E Mouse mAb (E6D7)

liquid, clone E6D7, Calbiochem®

Synonym(s):

Anti-ApoE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

E6D7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

should not react with

mouse, rabbit

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOE(348)

General description

Anti-Apolipoprotein E, mouse monoclonal, clone E6D7, recognizes the ~35 kDa Apo E2, E3, and E4 in human serum and brain. It is validated for ELISA, WB, IP, and paraffin and frozen sections.
Mouse monoclonal antibody supplied as diluted culture supernatant. Recognizes the ~35 kDa E2, E3, and E4 isoforms of apolipoprotein E.
Recognizes the ~35 kDa E2, E3, and E4 isoforms of apolipoprotein E in human serum and brain.

Immunogen

Human
a synthetic peptide corresponding to amino acids surrounding the polymorphic amino acid position 158 of ApoE

Application




ELISA (see comments)
Immunoblotting (1:100-1:1000)
Immunoprecipitation (see comments)
Frozen Sections (1:100-1:10,000)
Paraffin Sections (1:100-1:10,000, formic acid or heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

Culture supernatant diluted in PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Brain tissue, serum

Other Notes

Golabek, A.A., et al. 1996. J. Biol. Chem.271, 10602.
Recognizes the ~35 kDa E2, E3, and E4 isoforms of apolipoprotein E in human serum, but multiple bands are seen when used with brain tissue samples. Staining of formalin-fixed paraffin-embedded tissues may be enhanced by the pre-treatment of tissue with 70% formic acid for 10-30 min at room temperature or by heat pre-treatment using citrate buffer, pH 6.0. This antibody has also been reported to work for immunoprecipitation and ELISA. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abraham Kuot et al.
Clinical & experimental ophthalmology, 47(8), 1028-1042 (2019-06-18)
Fuchs endothelial corneal dystrophy (FECD) is a progressive and potentially a sight threatening disease, and a common indication for corneal grafting in the elderly. Aberrant thickening of Descemet's membrane, formation of microscopic excrescences (guttae) and gradual loss of corneal endothelial
Dale J Christensen et al.
Journal of immunology (Baltimore, Md. : 1950), 186(4), 2535-2542 (2011-02-04)
The molecular mechanism by which apolipoprotein E (apoE) suppresses inflammatory cytokine and NO production is unknown. Using an affinity purification approach, we found that peptide mimetics of apoE, derived from its receptor binding domain residues 130-150, bound to the SET
Veena Theendakara et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(3), 685-700 (2016-01-23)
A major unanswered question in biology and medicine is the mechanism by which the product of the apolipoprotein E ε4 allele, the lipid-binding protein apolipoprotein E4 (ApoE4), plays a pivotal role in processes as disparate as Alzheimer's disease (AD; in
Jing Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(6), 7748-7757 (2019-03-22)
Aging and apolipoprotein E4 (ApoE4) can increase the risk of cognitive impairment and neurodegenerative disorders, including Alzheimer's disease (AD), and patients with type 2 diabetes mellitus are highly susceptible to cognitive dysfunction. Recent research has indicated that metformin, a prescribed
Xia Hu et al.
Frontiers in neuroanatomy, 11, 45-45 (2017-06-24)
Genetic variations in the vacuolar protein sorting 10 protein (Vps10p) family have been linked to Alzheimer's disease (AD). Here we demonstrate deposition of fragments from the Vps10p member sortilin at senile plaques (SPs) in aged and AD human cerebrum. Sortilin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service