Skip to Content
MilliporeSigma

MXSMCMC24

Milliflex® Pre-filled Media Cassettes

MacConkey Agar, for lactose-fermenting bacteria, pack of 48 tests, suitable for bioburden testing

Synonym(s):

MacConkey Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116129
NACRES:
NA.74
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

International standards

Quality Segment

sterility

aseptically filled

form

plate (pre-poured agar)

shelf life

12 mo.

feature

ready-to-use

packaging

cassette of 10 mL, pack of 48 tests

manufacturer/tradename

Milliflex Oasis®

color

clear dark red

input

sample type beer
sample type pharmaceutical(s)
sample type cosmetics
sample type bottled water

pH

7.1±0.2 (25.0 °C)

application(s)

bioburden testing, cosmetics
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System

shipped in

ambient

storage temp.

2-8°C

suitability

suitable for (suitable for solid cultivation), coliforms, lactose-fermenting bacteria

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. MacConkey Agar is a selective medium for isolation of coliforms and E. coli. Bile salts and crystal violet present in the media largely inhibit the growth of the gram-positive microbial flora. Lactose and the pH indicator neutral red are used to detect lactose degradation. It is useful in differentiating between the lactose fermenting and non-lactose fermenting bacteria and also aid in isolation of gram-negative bacteria.

Application

Milliflex® Cassette prefilled with MacConkey Agar is used in Bioburden testing water monitoring. and general microbiological analysis. It is widely used in pharmaceutical and cosmetic industry

Features and Benefits

Standard Methods for the Examination of Water and Wastewater, United-States Pharmacopeia, European Pharmacopeia

Preparation Note

MacConkey

Analysis Note

Target Microorganisms: Lactose fermenting bacteria and coliforms
Lactose fermenting organisms will appear red with or without a zone of precipitated bile. Non-lactose fermenters will appear as colorless to white colonies. Crystal violet and bile salts inhibits Gram-positive microorganisms, especially Enterococci and Staphylococci
QC organisms
E. coli (ATCC 25922)

P. mirabilis (ATCC 12453)

S. aureus (ATCC 6538) as negative control

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library