Skip to Content
MilliporeSigma

MXRPCSZ

Power cord, Power MIX, Switzerland, 250V, 10A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32010600
UNSPSC Code:
41103801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

250 V, 10 A

Quality Segment

compatibility

for use with MOBIUS® Power MIX - all sizes

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library