Skip to Content
MilliporeSigma

MXRMTR2K

Motor, Mobius® Power MIX 1000/2000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32010600
UNSPSC Code:
41103801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

AC/DC input

120 V, 60 Hz, 15 A (connects to control box), 230 V, 50 Hz, 10 A (connects to control box)

material

stainless steel (in compliance with material code 1.4301 ASTM® 304)

parameter

4-30 °C temperature, 55 °C max. temp. (132 °F) at 1 atm (surface)

cable L

12 ft (3.65 m)

weight

30.4 lb (13.8 kg)

power consumption

0.55 kW

compatibility

for use with MOBIUS® Power MIX 1000 and 2000

Features and Benefits

CSA, CE, UL

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
MXRMTR2K04054839159367