Skip to Content
MilliporeSigma

MXPESP50R

Silicone O-ring + Plug Seal

Accessory for On-line sampling and filtration of aqueous liquids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
eCl@ss:
32160606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

silicone

Quality Segment

packaging

pkg of 5 numbers

manufacturer/tradename

MicropreSURE®

input

sample type pharmaceutical(s)
sample type dairy
sample type beverage(s)
sample type cider
sample type wine
sample type sport drink
sample type beer
sample type bottled water
sample type soft drink
sample type food(s)

application(s)

bioburden testing
food and beverages
pharmaceutical

shipped in

ambient

General description

Silicone O-ring + plug seal is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The Silicone O-ring + Plug Seal aids in the sealed and leak-free flow of the sample.

Application

Silicone O-ring + plug seal is used for in-process sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library