Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
23201100
form
beads
bead size
4-8 mesh
color
tan
shipped in
ambient
storage temp.
room temp
General description
4-8 Mesh Beads
Application
4-8 Mesh Beads
for Drying
Analysis Note
Color: Tan
Form: Beads
Identification (potassium): To pass test
Mesh size (4-8): 80% min
Form: Beads
Identification (potassium): To pass test
Mesh size (4-8): 80% min
Still not finding the right product?
Explore all of our products under Molecular Sieve, Type 3A
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MX1583C-4 | 04061839430570 |
