Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MTH17MAG-47K

Millipore

MILLIPLEX® Mouse TH17 Magnetic Bead Panel - Immunology Multiplex Assay

Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Luminex® Mouse Immunology Panel, Millipore Mouse Immunology Panel, Mouse Immunology Protein Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

linearity: 111.0%
(1:04)

linearity: 113.1%
(1:08)

standard curve range: 1.5-1,500 pg/mL
(IL-4)

standard curve range: 10-10,000 pg/mL
(IL-17F)

standard curve range: 127-130,000 pg/mL
(IL-28B)

standard curve range: 15-15,000 pg/mL
(IL-1β)

standard curve range: 2.4-2,500 pg/mL
(IL-22)

standard curve range: 20-20,000 pg/mL
(IL-10, IL-12(p70), & IL-21)

standard curve range: 3.4-3,500 pg/mL
(TNFα)

standard curve range: 34-35,000 pg/mL
(GM-CSF & IL-15)

standard curve range: 342-350,000 pg/mL
(IL-23)

standard curve range: 39-40,000 pg/mL
(IL-13 & IL-17A)

standard curve range: 4.9-5,000 pg/mL
(IL-5)

standard curve range: 488-500,000 pg/mL
(TNFβ)

standard curve range: 49-50,000 pg/mL
(CD40L IL-31 & MIP-3α)

standard curve range: 586-600,000 pg/mL
(IL-17E/ IL-25)

standard curve range: 6.9-6,000 pg/mL
(IL-2)

standard curve range: 7.8-8,000 pg/mL
(IFNγ & IL-6)

standard curve range: 78-80,000 pg/mL
(IL-33)

standard curve range: 879-900,000 pg/mL
(IL-27)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MILLIPLEX® Mouse Th17 Bead panel enables you to focus on the therapeutic potential of the modulation of CD4 T-helper cell cytokines. Coupled with the Luminex® xMAP®platform in a bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of dozens of analytes simultaneously, which can dramatically improve productivity.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There was no or minimum cross-reactivity between the antibodies and any other analytes in the panel (see details in assay protocol).

Application

  • Analytes: CD40 Ligand, GM-CSF, IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-21, IL-22, IL-23, IL-27, IL-28B, IL-31, IL-33, MIP-3α/CCL20, TNF-α, TNFβ
  • Recommended Sample type: serum, plasma or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: neat
  • Assay Run Time: overnight or one day
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Afzal Khan et al.
Cells, 10(5) (2021-06-03)
Interleukin-10 plays a vital role in maintaining peripheral immunotolerance and favors a regulatory immune milieu through the suppression of T effector cells. Inflammation-induced microvascular loss has been associated with airway epithelial injury, which is a key pathological source of graft
Mohammad Afzal Khan et al.
Frontiers in immunology, 9, 1010-1010 (2018-06-09)
Microvascular injury during acute rejection has been associated with massive infiltration of CD4+ T effector cells, and the formation of complement products (C3a and C5a). Regulatory T cells (Tregs) are potent immunosuppressors of the adaptive immune system and have proven
M John Albert et al.
Scientific reports, 9(1), 12024-12024 (2019-08-21)
There is no information on cytokine profiles for use as markers of protection in Campylobacter jejuni infection. To study this, we used outer membrane protein (MOMP [PorA]) as the vaccine for protection and spleen cell cytokines as markers of protection.
Sarah J Dulson et al.
Journal of immunology (Baltimore, Md. : 1950), 203(9), 2472-2484 (2019-09-29)
Innate lymphoid cells (ILCs) are strategically positioned at mucosal barrier surfaces where they respond quickly to infection or injury. Therefore, we hypothesized that ILCs are key contributors to the early immune response in the intestine against Listeria monocytogenes Using a
Anthony Tam et al.
Cells, 11(19) (2022-10-15)
Genome-wide association studies (GWAS) have shown that variants of patched homolog 1 (PTCH1) are associated with lung function abnormalities in the general population. It has also been shown that sonic hedgehog (SHH), an important ligand for PTCH1, is upregulated in

Related Content

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

MILLIPLEX® assays bridge immunology and cell signaling for inflammation studies, including T cell signaling and sepsis signaling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service