Skip to Content
MilliporeSigma

MR-107

EmbryoMax® KSOM Mouse Embryo Media

(1X), Liquid, with 1/2 Amino Acids w/o BSA

Synonym(s):

Embryonic stem cell media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Application

EmbryoMax® KSOM mouse embryo media has been used:
  • for culturing denuded mouse oocytes
  • in the washing step to perform mouse embryo collection
  • for culturing mouse zygotes

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library