Skip to Content

MR-025

EmbryoMax® FHM Mouse Embryo Media

(1X), Liquid, w/o Phenol Red

Synonym(s):

FHM Embryo Media, Mouse Embryo Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | embryo: suitable, cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Preparation Note

Can be stored at -20°C up to the expiration date on the bottle. Once thawed it should be used within 2 weeks

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under EmbryoMax® FHM Mouse Embryo Media


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library